No-Wire™ Autoclavable Test Tube Racks
No-Wire™ Autoclavable Test Tube Racks
Actual product may vary
Fisherbrand
No-Wire™ Autoclavable Test Tube Racks
Submersible polypropylene test tube racks sink in water baths and maintain stability even in agitated water. Great alternative to wire racks, with price and space saving advantages over other plastic racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPositions
FKS12662966Red40
FIS22-269954Blue60
FIS22-269979White40
FIS22-269948White60
FIS22-288141White21
FIS22-288145Blue21
FKS12652966Blue21
Catalog number FKS12662966
Price (SGD)
135.00
PK
Add to cart
Color:
Red
Positions:
40
Price (SGD)
135.00
PK
Add to cart
Submersible polypropylene test tube racks sink in water baths and maintain stability even in agitated water. Great alternative to wire racks, with price and space saving advantages over other plastic racks.
  • Suitable for use inside and outside the water bath to meet the demands of daily lab use
  • Heavier than water so the rack remains stable when submerged, even when agitated
  • Store flat to save space, but assembles quickly and easily
  • Features grids with numbered and lettered rows for tube identification
  • Interlocking feet facilitate easy stacking
  • Sturdy polypropylene is steam autoclavable at 121°C (250°F)
  • Round racks fit into 1000mL beaker and include removable, snap-on handle for easy retrieval
  • Rectangular racks measure 24.6 x 10.4 x 6.4cm (9.7 x 4.1 x 2.5 in.) and feature end plates for labeling
Specifications
Product TypeTube Rack
Array4 x 10
AutoclavableAutoclavable
ColorRed
HoldsTubes (16 to 20 mm)
MaterialPolypropylene
Positions40
ShapeRectangular
Shelves3 Tiers
StackableYes
SterilityNon-sterile
Dimensions (L x W x H)9.68 x 4.09 x 2.51 in. (24.59 x 10.4 x 6.4 cm)
Length (Metric)24.59 cm
Length (English)9.68 in.
Width (Metric)10.4 cm
Width (English)4.09 in.
Height (Metric)6.4 cm
Height (English)2.51 in.
Diameter (Metric) Opening20 mm
Unit SizePK