10μL Microvolume Filter Tips
10μL Microvolume Filter Tips
Actual product may vary
10μL Microvolume Filter Tips
Axygen Universal Tips designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
Have Questions?
Catalog NumberVolume (Metric)
FKS126595950.5 to 10 μL
Catalog number FKS12659595
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
0.5 to 10 μL
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention
Specifications
Volume (Metric)0.5 to 10 μL
FilteredFiltered
SterilityNon-sterile
ColorClear
Certifications/ComplianceUSP 3127
Clean ClaimsDNase, Rnase Free
For Use With (Application)Allows recovery of costly samples with improved precision and accuracy
FormatRack
Tip StyleMicro Point
Packaging96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
For Use With (Equipment)Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2