Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper
Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper
Store reagents, analytical standards and culture media in these glass reagent bottles. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper features excellent chemical resistance and can withstand high temperatures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS12653292250 mL
FKS12643292100 mL
FKS113378645000 mL
Catalog number FKS12653292
Price (SGD)
1,596.00
PK
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
1,596.00
PK
Add to cart
  • Complies with ISO 4796-2
  • Supplied with solid glass stopper
  • Pyrex™ borosilicate glass with ground neck
  • Autoclavable
Specifications
Capacity (Metric)250 mL
Diameter (Metric) Inner Neck19 mm
MaterialBorosilicate Glass
No. per Pack10/PK
Certifications/ComplianceISO 4796-2
Closure IncludedYes
MouthNarrow
Height (Metric)143 mm
Closure MaterialGlass
Closure Size19/26 mm
Diameter (Metric)73 mm
Closure TypeStopper
Unit SizePK