Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Size | Height (Metric) | Tolerance |
|---|---|---|---|---|
| FKS12653022 | 100 mL | 14/23 | 170 mm | ±0.2 mL |
| FKS12054448 | 1000 mL | 24/29 | 300 mm | ±0.8 mL |
| FKS12050703 | 2000 mL | 29/32 | 370 mm | ±1.2 mL |
| FKS12633022 | 25 mL | 10/19 | 110 mm | ±0.08 mL |
| FKS12020703 | 500 mL | 19/26 | 260 mm | ±0.5 mL |