Offering both strength and durability with economical replacement visors available in acetate or polycarbonate. Sperian™ Honeywell™ Clearways Faceshields are lightweight and comfortable enough to wear all day.
Have Questions?
Catalog Number
Product Type
Lens Coating
Lens Material
FKS12652733
Visor
Uncoated
Acetate
Catalog number FKS12652733
Price (SGD)
47.00
EA
Add to cart
Product Type:
Visor
Lens Coating:
Uncoated
Lens Material:
Acetate
Price (SGD)
47.00
EA
Add to cart
Recommended for the agriculture, chemical and forestry industries
Protects against hazards including abrasion, airborne particles, biohazards, chemicals, impacts, splashes and more
Safe, practical, economical integral visor fitting ensures extra protection