Check Order Status
Tissue Grinder
Actual product may vary
Fisherbrand
Tissue Grinder
Fits 1.5 and 2.0mL centrifuge tubes
Have Questions?
Catalog Number
Packaging
FKS12649595
Individually wrapped
Catalog number
FKS12649595
Price (SGD)
370.00
PK
Add to cart
Packaging:
Individually wrapped
Price (SGD)
370.00
PK
Add to cart
Durable and inert polypropylene construction
Designed to fit 1.5 mL and 2.0 mL microcentrifuge tubes
Specifications
Autoclavable
Autoclavable
Color
Blue
For Use With (Equipment)
1.5 and 2 mL microcentrifuge tube and PCR tube
Packaging
Individually wrapped
Material
Polypropylene
Sterility
Sterile