B-Line BL15 Clear Goggles
B-Line BL15 Clear Goggles
Actual product may vary
Fisherbrand
B-Line BL15 Clear Goggles
B-LINE is a range of safety eyewear accessible to all and is adapted to the most common industrial risks.
Have Questions?
Catalog NumberLens Coating
FKS12649130Uncoated
Catalog number FKS12649130
Price (SGD)
9.00
EA
Add to cart
Lens Coating:
Uncoated
Price (SGD)
9.00
EA
Add to cart
  • Clear translucent frame
  • Sealed or vented versions available
  • Adjustable strap
  • Fits over prescription glasses
Specifications
Lens CoatingUncoated
Lens TintClear
Product TypeSafety Goggles
Frame ColorClear
IncludesAdjustable strap
Weight (Metric)76 g
MaterialStrap: Nylon Polyester
Lens MaterialPolycarbonate
Frame MaterialPVC
Unit SizeEA