Bench Pipette Holder
Bench Pipette Holder
Actual product may vary
Bench Pipette Holder
Kartell™ Bench Pipette Holder is made of particularly clear polymethylmethacrylate (PMMA), which has high level finish that allows its content to be seen from any angle.
Have Questions?
Catalog NumberPositions
FKS126445664
Catalog number FKS12644566
Price (SGD)
1,088.00
EA
Add to cart
Positions:
4
Price (SGD)
1,088.00
EA
Add to cart
  • Complete with four tilted support shelves and white top
  • The four separate compartments are ideal for housing plastic or glass serologic pipettes of various capacities
  • Do not shine ultraviolet light on it
Specifications
ColorClear
HoldsPipets (1 to 50 mL)
MaterialPMMA
Positions4
For Use With (Equipment)Pipettes
Size290 x 85 x 89 mm Compartments
Dimensions (L x W x H)300 x 100 x 420mm
Unit SizeEA