Power Supply, Mini 300V Plus
Power Supply, Mini 300V Plus
Actual product may vary
Fisherbrand
Power Supply, Mini 300V Plus
Provides up to 300V (volts) and 400mA of power. Fisherbrand™ Power Supply, 300V Mini Plus includes dual voltage compatibility.
Have Questions?
Catalog Number
FKS12643546
Catalog number FKS12643546
Price (SGD)
1,824.00
EA
Add to cart
Price (SGD)
1,824.00
EA
Add to cart
  • Enhanced built-in safety features
  • Three digit LED display
  • Alarm function
  • Wipe clean polycarbonate housing