Upright Freezer Rack
Upright Freezer Rack
Actual product may vary
Fisherbrand
Upright Freezer Rack
Accommodates a variety of racks and storage boxes. Upright Freezer Rack includes side opening with handles on the front for easy box retrieval.
Have Questions?
Catalog NumberHolds
FKS12643476Tube Boxes
Catalog number FKS12643476
Price (SGD)
900.00
PK
Add to cart
Holds:
Tube Boxes
Price (SGD)
900.00
PK
Add to cart
  • Corrosion resistant stainless steel.
  • Suitable for use in low temperature storage freezers.
  • Holds 15 and 50 mL tube boxes.
Specifications
IncludesFront handle
Product TypeFreezer Rack
Array2 x 3
AutoclavableNot Autoclavable
ColorSilver
HoldsTube Boxes
MaterialStainless Steel
Positions1
ShapeRectangular
Shelves2 Tiers
StackableYes
SterilityNon-sterile
For Use With (Application)Tube box storage
For Use With (Equipment)15 and 50 mL Tube Boxes
Quantity1 Ea.
Dimensions (L x W x H)459 x 159 x 271 mm
Length (Metric)45.9 cm
Length (English)18.1 in.
Width (Metric)15.9 cm
Width (English)0.5 in.
Height (Metric)2.1 cm
Height (English)0.8 in.
Height (Metric) Interior134 mm
Unit SizePK