2890 Series Polycarbonate Lens Safety Goggles
2890 Series Polycarbonate Lens Safety Goggles
Actual product may vary
Fisherbrand
2890 Series Polycarbonate Lens Safety Goggles
Have Questions?
Catalog NumberLens CoatingVentilation Details
FKS12642605Anti-fog, Scratch-resistantSealed
Catalog number FKS12642605
Price (SGD)
65.00
EA
Add to cart
Lens Coating:
Anti-fog, Scratch-resistant
Ventilation Details:
Sealed
Price (SGD)
65.00
EA
Add to cart
Specifications
Lens CoatingAnti-fog, Scratch-resistant
Certifications/ComplianceEN 166:2001
Lens TintClear
Product TypeSafety Goggles
For Use With (Application)Construction,‎ laboratories
No. per Pack1 Ea.
Ventilation DetailsSealed
MaterialPolycarbonate
UV ProtectionYes
Unit SizeEA