Protective Coverall 4520, Hooded
Protective Coverall 4520, Hooded
Actual product may vary
Protective Coverall 4520, Hooded
Protective Coverall 4520, Hooded
Protective Coverall 4520, Hooded
Protective Coverall 4520, Hooded
Protective Coverall 4520, Hooded
Protective Coverall 4520, Hooded
Fisherbrand
Protective Coverall 4520, Hooded
Coveralls offer multiple layers of SMMMS for effective hold out and strength at a lightweight 43 gsm. Breathable and soft with a cloth like feel to be worn for extended periods to protect against static discharge, hazardous particulates, and liquid splash Type 5/6.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS12642565Large
3MT7000034761Medium
3MT7000034762Large
3MT7000034763X-Large
Catalog number FKS12642565
Price (SGD)
625.00
PK
Add to cart
Size:
Large
Price (SGD)
625.00
PK
Add to cart
  • Type 5/6 Coverall made from extremely lightweight and breathable material, designed to offer protection against hazardous dusts and limited liquid splashes and sprays
  • Made from super breathable SMMMS non-woven for effective release of body heat and moisture and reduced heat stress
  • Soft and strong multi-layered 43 gsm SMS for strong performance
  • 3 panel hood, 2-way zip and storm flap with elasticated back and ankles to reduce the risk of skin exposure
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out
  • Knitted cuffs for soft and secure arm placement
Specifications
Product TypeGeneral Purpose Coveralls
SizeLarge
GenderUnisex
MaterialPolypropylene
ColorWhite
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic, Knit
Neck StyleHooded
Hood Type3-panel
Closure TypeFront Zipper with Storm Flap
Seam TypeBound
SterilityNon-sterile
ResistanceDust, Liquid
CoatingAnti-static
Certifications/ComplianceCE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006
Certifications/Compliance (Europe)CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description3 Layers of Melt-blown Polypropylene in-between 2 layers of Spunbound Polypropylene
Latex FreeNo
Model4520
Size (English) Chest39 to 43 in.
Size (English) Inseam32 in.
Size (English) Waist51 in.
Size (Metric) Chest99.06 to 109.22 cm
Size (Metric) Inseam81.28 cm
Size (Metric) Waist129.54 cm
For Use With (Application)Insulation Installation, Asbestos and Lead Abatement, Coal Dust in Power Plants, Woodworking, Metal Polishing, Light-Duty Building Cleaning, General Powder Handling, General Industrial Clean-up and Maintenance, Machine or Vehicle Maintenance and Food Processing
Height (English)5.75 to 6 ft.
Height (Metric)174 to 181 cm
Unit SizePK