Check Order Status
Technical pH Buffer Solution, pH 9.21, Mettler Toledo™
Element Search
Structure Search
Fisher Catalogue
Technical pH Buffer Solution, pH 9.21, Mettler Toledo™
Traceable to the primary standards of reference materials selected by NIST.
Have Questions?
Catalog Number
Quantity
FKS12631306
6 x 250 mL
Catalog number
FKS12631306
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
6 x 250 mL
Request bulk or custom format
Specifications
Color
Blue
pH
9.21
Physical Form
Liquid
Quantity
6 x 250 mL
Solution Type
pH Buffer
View more
pH 9.21