Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Actual product may vary
Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Fisherbrand
Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Designed to deliver optimal results in wet or dry work environments where chemical resistance is crucial. Fully reusable, with an unequaled abrasion resistance, it provides superb comfort for the wearer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS126308626
FKS1240514010
Catalog number FKS12630862
Price (SGD)
-
Size:
6
  • Reversed lozenge finish further enhances levels of grip.
  • Longer length (38cm) than standard, extending protection to the wrist and lower forearm area
  • Flocked lining and flexible nitrile file offer exceptional comfort
  • Gauntlet style
  • Reusable
  • Latex free
  • Resistant to chemicals and liquid protection
  • CE, EN374-3:2003, CATEGORY III
Specifications
Back MaterialNitrile
Cuff MaterialNitrile
Exterior FinishSmooth
Product TypeImmersion Gloves
Size6
Style37-675
Thickness (Metric)0.38 mm
Glove Cuff ColorGreen
Glove Cuff StyleGauntlet
Liner MaterialFlocked
Quantity12 Pairs
Resistant ToChemical resistance
Series37-675
Unit SizePK