Stainless Steel Round Immersion Basket with Handles
Stainless Steel Round Immersion Basket with Handles
Actual product may vary
Fisherbrand
Stainless Steel Round Immersion Basket with Handles
Facilitates complete immersion within S 50 R Ultrasonic Cleaner baths. Fisherbrand™ Stainless Steel Round Immersion Baskets with Handles feature a functional and modern design.
Have Questions?
Catalog number FKS12629355
Price (SGD)
557.00
PK
Add to cart
Price (SGD)
557.00
PK
Add to cart
  • Mesh bottom
  • Two handles, one on each side
  • Handles are coated with plastic
Specifications
TypeBasket
Depth (Metric)575 mm
Diameter (Metric)220 mm
For Use With (Equipment)Ultrasonic Unit, Elmasonic S 50 R
Height (Metric)70 mm
Unit SizePK