Quickfit™ Weighing Bottles with Ground Glass Stoppers
Quickfit™ Weighing Bottles with Ground Glass Stoppers
Actual product may vary
Fisherbrand
Quickfit™ Weighing Bottles with Ground Glass Stoppers
Manufactured from chemically inert Pyrex™ Borosilicate Glass and complies with DIN 12605. Quickfit™ Weighing Bottles with Ground Glass Stoppers are available in a variety of capacities with choice of squat or tall form bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Form
FKS1262865010 mLSquat
FKS124010418 mLTall
FKS1241104120 mLTall
FKS1245104120 mLSquat
FKS1242104125 mLTall
FKS1243104140 mLTall
FKS1244104160 mLTall
FKS12461041100 mLTall
Catalog number FKS12628650
Price (SGD)
78.00
EA
Add to cart
Capacity (Metric):
10 mL
Form:
Squat
Price (SGD)
78.00
EA
Add to cart
Manufactured from chemically inert Pyrex™ Borosilicate Glass and complies with DIN 12605. Quickfit™ Weighing Bottles with Ground Glass Stoppers are available in a variety of capacities with choice of squat or tall form bottles.
  • Complies with DIN 12605
  • Chemically inert Pyrex™ Borosilicate Glass
  • Lightweight construction with ground glass stoppers
  • Choice of squat or tall form bottles
Specifications
Capacity (Metric)10 mL
FormSquat
IncludesGround glass stopper
MaterialBorosilicate Glass
Certifications/ComplianceDIN 12605
Closure IncludedYes
Height (Metric)30 mm
Closure MaterialGround Glass
Diameter (Metric)35 mm
Closure TypeStopper
Unit SizeEA