Polypropylene Scoop
Polypropylene Scoop
Actual product may vary
Polypropylene Scoop
Polypropylene Scoop
Fisherbrand
Polypropylene Scoop
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ Polypropylene Scoops are lighter than metal, don’t dent and feature good chemical resistance. Also suitable as weighing scoops.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)
FKS12623566250 mL260 mm
FKS1261356650 mL160 mm
Catalog number FKS12623566
Price (SGD)
83.00
PK
Add to cart
Capacity (Metric):
250 mL
Length (Metric):
260 mm
Price (SGD)
83.00
PK
Add to cart
  • White polypropylene scoop
  • With precision formed filling edge and comfortable, stable handle
  • Easily readable volume quantities on the upper side of the handle
Specifications
Product TypeMeasuring Scoop
MaterialPolypropylene
Capacity (Metric)250 mL
ColorWhite
For Use With (Application)For contact with food
Length (Metric)260 mm
Unit SizePK