Check Order Status
Pyrex™ Borosilicate Glass Hemolysis Test Tube
Fisherbrand
Pyrex™ Borosilicate Glass Hemolysis Test Tube
Have Questions?
Catalog Number
Diameter (Metric)
FKS12623092
12 mm
Catalog number
FKS12623092
Price (SGD)
179.00
PK
Add to cart
Diameter (Metric):
12 mm
Price (SGD)
179.00
PK
Add to cart
Specifications
Capacity (Metric)
4 mL
Length (Metric)
65 mm
Material
Borosilicate Glass
For Use With
For hemolysis
Quantity
100 Pack
Autoclavable
Autoclavable
Diameter (Metric)
12 mm