Universal Oven UF with Forced-Air Convection & SingleDISPLAY
Universal Oven UF with Forced-Air Convection & SingleDISPLAY
Actual product may vary
Universal Oven UF with Forced-Air Convection & SingleDISPLAY
Universal Oven UF with Forced-Air Convection & SingleDISPLAY
Fisherbrand
Universal Oven UF with Forced-Air Convection & SingleDISPLAY
Ideal for drying, heating, ageing, burn-in and hardening in research, science, industry and quality assurance.
Have Questions?
Catalog NumberElectrical RequirementsCapacity (Metric)
FKS12616997115/230 V, 50/60 Hz108 L
Catalog number FKS12616997
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Electrical Requirements:
115/230 V, 50/60 Hz
Capacity (Metric):
108 L
Precise, easy to use, high quality - these three words describe the Memmert Universal Oven UF.
  • Model sizes: 30, 55, 75, 110, 160, 260, 450, 750, 1060
  • Temperature adjustment range: up to +300°C
  • Defined and programme-controlled fan speed
  • All-round heating system: absolutely uniform and precise temperature distribution
  • Pre-heated fresh air prevents temperature fluctuations
  • AtmoCONTROL software on a USB stick for programming, managing and transferring programmes via Ethernet interface or USB port
  • ControlCOCKPIT: SingleDISPLAY - Adaptive multifunctional digital PID-microprocessor controller with high-definition TFT-colour display
  • adjustable parameters: temperature (Celsius or Fahrenheit), fan speed, air flap position, programme time, time zones, summertime/wintertime
  • Overtemperature protection
The technologically perfected masterpiece made of high-quality, hygienic, easy-to-clean stainless steel leaves nothing to be desired in terms of ventilation and control technology.
Specifications
Capacity (English)3.8 cu. ft.
Certifications/ComplianceCE, cUL, DVE, EAC, GS
Chamber MaterialStainless Steel (304/1.4301)
Controller TypeSingleDISPLAY. Adaptive multifunctional digital PID-microprocessor controller
Convection TypeForced
Capacity (Metric)108 L
Temperature (Metric)20°C to 300°C
TypeDrying Oven
Display1 high-definition TFT-colour display (SingleDISPLAY)
For Use With (Application)Temperature Control in Science, Research and Material Tests in Industry
InsulationFully isolated Stainless Steel door with 2-point locking (compression door lock)
MaterialStainless Steel
Max. Shelf Load20 kg
ModelUF110
No. of Shelves5
Plug TypeMains cable with plug (German type)
Safety FeaturesTemperature control, autodiagnostic system
Shelf MaterialStainless Steel
Temperature ControlAdaptive multifunctional digital PID-microprocessor controller
Temperature StabilityUp to 99.9°C: 0.1 / from 100°C: 0.5
TimerDigital backwards counter with target time setting, adjustable from 1 minute to 99 days
Warranty3 years
Dimensions (D x W x H) Interior400 x 560 x 480 mm
Dimensions (L x W x H)745 x 864 x 584 mm
Length (Metric)584 mm
Width (Metric)745 mm
Width (Metric) Interior560 mm
Height (Metric)864 mm
Height (Metric) Interior480 mm
Depth (Metric)584 mm
Depth (Metric) Interior400 mm
Shipping Weight (Metric)103 kg
Weight (Metric)74 kg
Electrical Requirements115/230 V, 50/60 Hz
Voltage115/230 V
Wattage1800/2800 W