Check Order Status
Lamp Stands
Actual product may vary
Fisherbrand
Lamp Stands
Allows easy and comfortable hands free operation of UV lamps
Have Questions?
Change view
Catalog Number
Model
FKS12611732
J-138
FKS11786812
J-129
2 Options
Catalog number
FKS12611732
Price (SGD)
925.00
EA
Add to cart
Model:
J-138
Price (SGD)
925.00
EA
Add to cart
Allows easy and comfortable hands free operation of UV lamps
Housing: Varies from metal housing with scratch resistant powder coat to plastic housing
Allows hands free operation of the lamp
Specifications
Dimensions (L x W x H)
13 x 3 x 10.74 in. (330 x 76 x 273 mm)
For Use With (Equipment)
EL-series UV lamp
Height (English)
10.74 in.
Height (Metric)
273 mm
Length (English)
13 in.
Length (Metric)
330 mm
Model
J-138
Packaging Quantity
1 Ea.
Width (English)
3 in.
Width (Metric)
76 mm
Product Type
Lamp Stand