Azlon™ Polypropylene Wide Mouth Containers with Screw Caps
Azlon™ Polypropylene Wide Mouth Containers with Screw Caps
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Wide Mouth Containers with Screw Caps
Manufactured from Polypropylene. Azlon™ Polypropylene Wide Mouth Containers with Screw Caps are autoclavable with unlined screw caps. Ideal for storage of samples, powders, animal feeds, and much more.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FKS12594997250 mL36/PK
FKS12545007950 mL24/PK
Catalog number FKS12594997
Price (SGD)
459.00
PK
Add to cart
Capacity (Metric):
250 mL
Quantity:
36/PK
Price (SGD)
459.00
PK
Add to cart
Manufactured from Polypropylene. Azlon™ Polypropylene Wide Mouth Containers with Screw Caps are autoclavable with unlined screw caps. Ideal for storage of samples, powders, animal feeds, and much more.
  • Autoclavable wide mouth containers with unlined screw caps
  • Good chemical resistance
  • Ideal for storage of samples, powders, animal feeds, and much more.
Specifications
Capacity (Metric)250 mL
For Use With (Application)Storage of Samples, Powders, Animal Feeds
IncludesUnlined Screw Cap
MaterialPolypropylene
Quantity36/PK
Temperature Range (Metric)–20 to 135°C
Weight (Metric)2.05 kg
ClarityTranslucent
Closure IncludedYes
AutoclavableAutoclavable
MouthWide
ShapeRound
Height (Metric)66 mm
Product TypeStorage Container
Closure LinerUnlined
Diameter (Metric)89 mm
Closure TypeScrew Cap
Unit SizePK