Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Actual product may vary
Fisherbrand
Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) OuterClosure Size
FKS125888662000 mL280 mm166 mm42
FKS11960905500 mL160 mm105 mm42
Catalog number FKS12588866
Price (SGD)
1,171.00
PK
Add to cart
Capacity (Metric):
2000 mL
Height (Metric):
280 mm
Diameter (Metric) Outer:
166 mm
Closure Size:
42
Price (SGD)
1,171.00
PK
Add to cart
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
  • PBT screw caps with PTFE faced sealing disc
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ Borosilicate Glass
  • Highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
  • Graduated to full capacity
Specifications
Closure MaterialSVL™ Phenolic, PTFE-faced with rubber sealing disc
Capacity (Metric)2000 mL
MaterialBorosilicate Glass, Borosilicate Glass
Height (Metric)280 mm
IncludesApproximate graduations
ShapeConical
GraduatedYes
Closure TypeScrew Cap
Diameter (Metric) Outer166 mm
Closure Size42
Unit SizePK