Azlon™ Tapered Containers with Snap-On Lid
Azlon™ Tapered Containers with Snap-On Lid
Actual product may vary
Fisherbrand
Azlon™ Tapered Containers with Snap-On Lid
Constructed of heavy duty, chemically resistant Polypropylene and autoclavable at 121°C Azlon™ Tapered Containers with Snap-On Lid includes a moulded handle for safe and secure carrying. Squat form provides easy location on shelves, bench top, or while in transit. Moulded graduations add quick, easy volume assessment. Caps on 10 & 20 litre carboys have moulded drill guides to aid installation of feed-lines.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Includes
FKS125850872500 mLTight Fitting LDPE Lid
FKS120169022000 mLTight Fitting LDPE Lid
Catalog number FKS12585087
Price (SGD)
404.00
PK
Add to cart
Capacity (Metric):
2500 mL
Includes:
Tight Fitting LDPE Lid
Price (SGD)
404.00
PK
Add to cart
  • Useful lightweight tapered containers with tight-fitting LDPE lids
  • Good chemical resistance
  • Ideal for storage of samples, powders, animal feeds, and more
  • Suitable for food contact applications in accordance with USA regulations 
Specifications
Capacity (Metric)2500 mL
For Use With (Application)Storage of Samples, Powders, Animal Feeds
IncludesTight Fitting LDPE Lid
MaterialHDPE
Quantity25/PK
Temperature Range (Metric)–20 to 135°C
Weight (Metric)2.82 kg
Closure IncludedYes
AutoclavableAutoclavable
MouthWide
ShapeRound
Height (Metric)150 mm
Product TypeStorage Container
Closure MaterialLDPE
Diameter (Metric)170 mm
Closure TypeSnap Lid
Unit SizePK