Pyrex™ Short Stem Glass Funnel
Pyrex™ Short Stem Glass Funnel
Actual product may vary
Fisherbrand
Pyrex™ Short Stem Glass Funnel
Manufactured from chemical and heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Short Stem Glass Funnel with 60° bowl angle provides precise fitting of filter paper.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopHeight (Metric)
FKS1258506735mm60 mm
FKS1252994570mm125 mm
FKS11397934150mm270 mm
FKS1134793445mm78 mm
FKS11992568105mm186 mm
Catalog number FKS12585067
Price (SGD)
618.00
PK
Add to cart
Diameter (Metric)Top:
35mm
Height (Metric):
60 mm
Price (SGD)
618.00
PK
Add to cart
Manufactured from chemical and heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Short Stem Glass Funnel with 60° bowl angle provides precise fitting of filter paper.
  • With 60° bowl angle for precise fitting of filter paper
  • Manufactured from chemical and heat resistant Pyrex™ borosilicate glass
Specifications
Diameter (Metric)Top35mm
Diameter (Metric) Stem8mm
MaterialPyrex Glass, Borosilicate Glass
Height (Metric)60 mm
Certifications/ComplianceNF B 35 032
Diameter (Metric) Disc45 to 55 mm (Filter Paper)
Length (Metric) Stem35mm
Product TypeShort stem funnel
Unit SizePK