Foam Ear Plugs
Foam Ear Plugs
Actual product may vary
Fisherbrand
Foam Ear Plugs
Foam ear plugs are an economical choice for disposable hearing protection. Hypoallergenic and a tapered design help provide a noise-reducing seal in the ear canal.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelCorded
FKS12580147K1110Corded
FIS19-0720761100Uncorded
Catalog number FKS12580147
Price (SGD)
191.00
PK
Add to cart
Model:
K1110
Corded:
Corded
Price (SGD)
191.00
PK
Add to cart
  • Disposable orange ear plugs
  • Made from hypoallergenic material for maximum protection and comfort
  • Smooth, dirt-resistant surface for hygiene
  • Recommended for high noise situations (up to 100 dBA TWA)
Specifications
SizeUniversal
For Use With (Application)Assembly, blasting, cleaning, demolition, electrical, facility maintenance, grinding, machine operations, painting, sanding, welding
ModelK1110
Pairs per Package100 Pair
Noise Reduction Rating (NRR)29 dB
CordedCorded
Plug ShapeTapered