Check Order Status
Quickfit™ Borosilicate Glass Medium Neck Round Bottom Flask
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Medium Neck Round Bottom Flask
Complies with BS 2735
Have Questions?
Catalog Number
Capacity (Metric)
Height (Metric)
FKS12578895
250 mL
165 mm
Catalog number
FKS12578895
Price (SGD)
230.00
PK
Add to cart
Capacity (Metric):
250 mL
Height (Metric):
165 mm
Price (SGD)
230.00
PK
Add to cart
Complies with BS 2735
With a longer neck than standard Quickfit™ flasks
Useful for reducing problems associated with frothing and splashing
Specifications
Capacity (Metric)
250 mL
Material
Borosilicate Glass
For Use With (Application)
Useful for reducing problems associated with frothing and splashing
Height (Metric)
165 mm
Neck Style
Medium
Socket Joint
24/29