Quickfit™ Borosilicate Glass Liebig Condenser
Quickfit™ Borosilicate Glass Liebig Condenser
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Liebig Condenser
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS12575057400 mm
FKS12019652260 mm
FKS12039652390 mm
FKS12049652530 mm
Catalog number FKS12575057
Price (SGD)
433.00
PK
Add to cart
Length (Metric):
400 mm
Price (SGD)
433.00
PK
Add to cart
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
  • Complies with DIN 12576
  • Popular water-jacketed condenser suitable for distillation and reflux applications
  • With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
Specifications
Certifications/ComplianceDIN 12576
For Use With (Application)Suitable for distillation and reflux applications
Length (Metric)400 mm
Standard Taper Joint No29/32
MaterialBorosilicate Glass
TypeLiebig Condenser
Unit SizePK