PTFE Screwthread Connector
Product Image
Fisherbrand
PTFE Screwthread Connector
Attaches easily to glassware with flexible tubing. Pyrex™ PTFE Screwthread Connectors are ideal for connections in contact with aggressive liquids or chemical vapours.
Have Questions?
Catalog NumberMaterial
FKS12571246PTFE
Catalog number FKS12571246
Price (SGD)
474.00
PK
Add to cart
Material:
PTFE
Price (SGD)
474.00
PK
Add to cart
Attaches easily to glassware with flexible tubing. Pyrex™ PTFE Screwthread Connectors are ideal for connections in contact with aggressive liquids or chemical vapours.
  • For use with glassware manufactured by SciLabware Ltd having 13mm screwthread inlets and outlets
  • Allows 9mm bore rubber hose and flexible tubing to be attached to the glassware easily and safely
  • PTFE protected 'O' rings for use up to 200°C
  • High chemical resistance
Specifications
MaterialPTFE
For Use With9mm bore
FlexibilityFlexible
Max. Operating Temperature (Metric)200°C
Product TypeTubing Connector
Unit SizePK