Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm
Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm
Manufactured from Borosilicate Glass and complies with ISO 6556. Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm features heavy walls to provide increased mechanical strength while 'studded' base minimises surface abrasion. These Büchner type flasks include an integral glass side-arm to accept 9mm bore flexible tubing necessary for vacuum filtration.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) Inner NeckDiameter (Metric) Outer
FKS125599653000 mL335 mm68 mm187 mm
FKS11941548250 mL155 mm34 mm85 mm
FKS120898972000 mL260 mm60 mm166 mm
FKS113878345 L385 mm68 mm220 mm
Catalog number FKS12559965
Price (SGD)
3,817.00
PK
Add to cart
Capacity (Metric):
3000 mL
Height (Metric):
335 mm
Diameter (Metric) Inner Neck:
68 mm
Diameter (Metric) Outer:
187 mm
Price (SGD)
3,817.00
PK
Add to cart
Manufactured from Borosilicate Glass and complies with ISO 6556. Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm features heavy walls to provide increased mechanical strength while 'studded' base minimises surface abrasion. These Büchner type flasks include an integral glass side-arm to accept 9mm bore flexible tubing necessary for vacuum filtration.
  • Complies with ISO 6556
  • Büchner type flasks for vacuum filtration
  • Heavy walls provide increased mechanical strength
  • Studded base to minimise surface abrasion
  • With integral glass sidearm to accept 9mm bore flexible tubing
Specifications
Certifications/ComplianceISO 6556
DescriptionStyle: Erlenmeyer
Diameter (Metric) Inner Neck68 mm
Capacity (Metric)3000 mL
MaterialBorosilicate Glass
Height (Metric)335 mm
Diameter (Metric) Outer187 mm
Unit SizePK