Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes
Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes
Medium walled test tubes with rimmed edgeManufactured from Pyrex™ borosilicate glass to give superior performance over neutral borosilicate and soda lime glass and excellent chemical resistance. Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes are ideal for microbiology culture work.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FKS1255947524 mm200 mm73 mL
FKS1252946512 mm100 mm8 mL
Catalog number FKS12559475
Price (SGD)
486.00
PK
Add to cart
Diameter (Metric):
24 mm
Length (Metric):
200 mm
Capacity (Metric):
73 mL
Price (SGD)
486.00
PK
Add to cart
Manufactured from Pyrex™ borosilicate glass to give superior performance over neutral borosilicate and soda lime glass and excellent chemical resistance. Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes are ideal for microbiology culture work.
  • Rimmed medium walls: constructed for optimum performance
  • Robust construction: allows use at high temperatures up to 500deg.C
  • Conforms to: ISO 4142
Specifications
Certifications/ComplianceISO 4142
Capacity (Metric)73 mL
Length (Metric)200 mm
MaterialBorosilicate Glass
Quantity50 Pack
RimmedYes
Diameter (Metric)24 mm
Unit SizePK