Pyrex™ Büchner Flask with Screw Thread Connector
Pyrex™ Büchner Flask with Screw Thread Connector
Actual product may vary
Fisherbrand
Pyrex™ Büchner Flask with Screw Thread Connector
Manufactured from borosilicate glass. Pyrex™ Büchner Flask with Screw Thread Connector features heavy wall construction for vacuum filtration applications, and tooled neck to accept a drilled rubber stopper. Sidearm includes a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing. Complies with ISO 6556.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) Inner TubingDiameter (Metric) Outer
FKS125591251000 mL230 mm45 mm (Neck I.D.)135 mm
FKS113978345 L385 mm68 mm (Neck I.D.)220 mm
Catalog number FKS12559125
Price (SGD)
1,169.00
PK
Add to cart
Capacity (Metric):
1000 mL
Height (Metric):
230 mm
Diameter (Metric) Inner Tubing:
45 mm (Neck I.D.)
Diameter (Metric) Outer:
135 mm
Price (SGD)
1,169.00
PK
Add to cart
Manufactured from borosilicate glass. Pyrex™ Büchner Flask with Screw Thread Connector features heavy wall construction for vacuum filtration applications, and tooled neck to accept a drilled rubber stopper. Sidearm includes a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing. Complies with ISO 6556.
  • Complies with ISO 6556
  • Büchner flasks with heavy wall construction for vacuum filtration applications
  • Sidearm features a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing
  • With tooled neck to accept a drilled rubber stopper
Specifications
Certifications/ComplianceISO 6556, DIN 12476
Diameter (Metric) Inner Tubing45 mm (Neck I.D.)
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
Height (Metric)230 mm
IncludesPTFE screw thread connector
Diameter (Metric) Outer135 mm
Unit SizePK