Shandon™ Tissue-Loc™ Tissue Cassettes IV
Shandon™ Tissue-Loc™ Tissue Cassettes IV
Actual product may vary
Fisherbrand
Shandon™ Tissue-Loc™ Tissue Cassettes IV
Compatible with Epredia™ Shandon™ MicroWriter™ and PrintMate™ cassette labelers
Have Questions?
Catalog NumberColor
FKS12550077White
Catalog number FKS12550077
Price (SGD)
-
Color:
White
  • Disposable
  • Holds tissue specimens during embedding, processing
  • Pore style: Small slots
  • Supplied in box of 250 cassettes with attached lids
Specifications
Pore StyleSmall Slots
Closure TypeHinged