200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference
200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference
Actual product may vary
Fisher Catalogue
200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference
Designed to fit a wide variety of single and multi-channel pipettes
Have Questions?
Catalog NumberSterilityColorFormat
FKS12545547Non-sterileYellowRack
Catalog number FKS12545547
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Sterility:
Non-sterile
Color:
Yellow
Format:
Rack
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
Specifications
Volume (Metric)1 to 200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorYellow
AutoclavableAutoclavable
Clean ClaimsDNase, RNase, and Pyrogen free
FormatRack
Tip StyleBeveled, Universal
PackagingHinged rack
For Use With (Equipment)Universal Fit
GraduatedYes
MaterialPolypropylene
Graduations10, 50, 100μL
Unit SizePK