Pyrex™ Crystallizing Dish with Spout
Pyrex™ Crystallizing Dish with Spout
Actual product may vary
Fisherbrand
Pyrex™ Crystallizing Dish with Spout
Manufactured from Pyrex™ borosilicate glass for excellent chemical and thermal resistance. Pyrex™ Borosilicate Crystallizing Dishes with Spout provide a superior device for maximum evaporation to occur.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS12540436500 mL115 mm65 mm
Catalog number FKS12540436
Price (SGD)
1,007.00
PK
Add to cart
Capacity (Metric):
500 mL
Diameter (Metric):
115 mm
Height (Metric):
65 mm
Price (SGD)
1,007.00
PK
Add to cart
  • Integral spout for excess liquid
  • Manufactured from Pyrex borosilicate glass
  • Thermal resistant to 400°C
  • Complies with DIN 12338
Specifications
Certifications/ComplianceDIN 12337
Bottom ShapeFlat
Capacity (Metric)500 mL
MaterialBorosilicate Glass
ShapeRound
TypeCrystallizing Dish
Height (Metric)65 mm
Diameter (Metric)115 mm
Unit SizePK