LDPE DIY Printed Safety Wash Bottles
LDPE DIY Printed Safety Wash Bottles
Actual product may vary
LDPE DIY Printed Safety Wash Bottles
LDPE DIY Printed Safety Wash Bottles
Fisher Catalogue
LDPE DIY Printed Safety Wash Bottles
Create custom labels for these do-it-yourself LDPE wash bottles. Azlon™ LDPE DIY Printed Safety Wash Bottles come with adhesive hazard labels and are ideal for a variety of liquid storage and sampling applications. DripLok™ vapor venting caps provide increased safety with volatile solvents.
Have Questions?
Catalog NumberCapacity (Metric)Closure TypeClosure Color
FKS12540066250 mLVapor-ventingWhite
Catalog number FKS12540066
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Closure Type:
Vapor-venting
Closure Color:
White
Create custom labels for these do-it-yourself LDPE wash bottles. Azlon™ LDPE DIY Printed Safety Wash Bottles come with adhesive hazard labels and are ideal for a variety of liquid storage and sampling applications. DripLok™ vapor venting caps provide increased safety with volatile solvents.
  • Do-it-yourself wash bottles with blank write-on panel
  • Indelibly printed in three languages (English, French and German)
  • Supplied with adhesive hazard labels
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorTranslucent
Diameter (Metric) Outer Neck38 mm
Diameter (Metric) Inner Neck34 mm
IncludesClosure, blank write-on panel, adhesive hazard signs
MaterialLDPE
Quantity5/PK
Temperature Range (Metric)–50 to 80°C
Closure ColorWhite
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthWide
ShapeCylindrical
Height (Metric)140 mm
Product TypeDIY Printed Safety Wash Bottle
Closure MaterialPolypropylene
Diameter (Metric)60 mm
Closure TypeVapor-venting
Through the Closure DispensingYes
Unit SizePK