Presterilized Conical Centrifuge Tubes
Presterilized Conical Centrifuge Tubes
Actual product may vary
Fisherbrand
Presterilized Conical Centrifuge Tubes
Graduated tubes with conical bottom
Have Questions?
Catalog NumberCapacity (Metric)Packaging
FKS1253910750 mL25 Tubes and Caps/Rack, 20 Racks/Cs.
Catalog number FKS12539107
Price (SGD)
889.00
PK
Add to cart
Capacity (Metric):
50 mL
Packaging:
25 Tubes and Caps/Rack, 20 Racks/Cs.
Price (SGD)
889.00
PK
Add to cart
  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
Specifications
Cap StyleScrew Cap
Closure MaterialHDPE
AutoclavableNot Autoclavable
Capacity (Metric)50 mL
ColorClear
MaterialPolypropylene
For Use With (Application)For sample identification
IncludesScrew cap
No. per Pack250/Pk.
Packaging25 Tubes and Caps/Rack, 20 Racks/Cs.
SterilitySterile
GraduatedYes
Closure TypeScrew Cap
Bottom ShapeCylindrical
Max. RCF12,000 x g
Product TypeCentrifuge Tube
Unit SizePK