Azlon™ Class A PMP Volumetric Flask
Azlon™ Class A PMP Volumetric Flask
Actual product may vary
Fisherbrand
Azlon™ Class A PMP Volumetric Flask
Complete with printed batch code. The Azlon Class A PMP Volumetric Flask includes a polypropylene stopper.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS12530076500 mL
FKS1250007650 mL
Catalog number FKS12530076
Price (SGD)
203.00
EA
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
203.00
EA
Add to cart
Complete with printed batch code. The Azlon Class A PMP Volumetric Flask includes a polypropylene stopper.
  • Manufactured from glass clear PMP offering excellent clarity
  • Individually adjusted ring mark to class A tolerances according to ISO 1042
  • Complete with printed batch code
  • Batch certificate available to download online
  • Supplied with polypropylene stopper
Specifications
Certifications/ComplianceISO 1042
Closure MaterialPolypropylene
ColorClear
Capacity (Metric)500 mL
MaterialPMP
Height (Metric)270 mm
IncludesStopper
No. per Pack1
Temperature Range (Metric)-20°C to +135°C
Tolerance±0.25 mL
AutoclavableAutoclavable
ClassA
Closure TypeStopper
Unit SizeEA