Innova™ 40R Incubator Shaker
Innova™ 40R Incubator Shaker
Actual product may vary
Fisherbrand
Innova™ 40R Incubator Shaker
Use these versatile incubator shakers for support high-speed applications and heavy workloads. New Brunswick Scientific™ Innova™ 40R Incubator Shakers provide very broad temperature capabilities for culturing a wide variety of organisms in flasks up to 3 liters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberOrbit
FKS1252888625 mm
FKS1251888619 mm
Catalog number FKS12528886
Price (SGD)
67,771.00
EA
Add to cart
Orbit:
25 mm
Price (SGD)
67,771.00
EA
Add to cart
  • Exceptionally quiet operation provides a favorable work environment
  • Triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble free operation for years
  • Compact benchtop model – fits easily on the bench and holds flasks up to 3L
  • Clear cover allows easy viewing of the cultures and lifts easily to access all samples
  • Versatile accessory platform (sold separately) accommodates flasks up to 3L as well as test tube racks and Microplate holders
  • Certified. Meets cETL, UL, CSA and CE standards
Specifications
Orbit25 mm
Speed Range25 to 500 rpm
Electrical Requirements230 V, 50 Hz
Voltage230 V
Holds3 L
Product TypeIncubator Shaker
Frequency50 Hz
Depth (Metric)76.2  cm
Height (Metric)101.6 cm
Width (Metric)55.9 cm
ModelInnova 40R
Timer Range0.01 to 99:59 hr.
Unit SizeEA