Azlon™ Polypropylene Microcentrifuge Tube Rack
Azlon™ Polypropylene Microcentrifuge Tube Rack
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Microcentrifuge Tube Rack
Economical and sturdy test tube racks supplied in self assembly flat packs
Have Questions?
Catalog NumberColor
FKS12521406Blue
Catalog number FKS12521406
Price (SGD)
448.00
PK
Add to cart
Color:
Blue
Price (SGD)
448.00
PK
Add to cart
  • Hole size 11mm square
  • Accepts 128 x 1.5mL microcentrifuge tubes
  • Autoclavable at 121°C
  • Sinks in water baths
  • Alpha numeric place coding
Specifications
DescriptionMicrocentrifuge tube rack
AutoclavableAutoclavable
ColorBlue
MaterialPolypropylene
Positions128
For Use With (Equipment)Microcentrifuge tubes
Dimensions (L x W x H)265 x 126 x 38 mm
Length (Metric)265 mm
Length (English)10.4 in.
Width (Metric)126 mm
Width (English)4.96 in.
Height (Metric)38 mm
Height (English)1.49 in.
Unit SizePK