PYREX™ Heavy-Duty Griffin Beakers
PYREX™ Heavy-Duty Griffin Beakers
Actual product may vary
Fisher Catalogue
PYREX™ Heavy-Duty Griffin Beakers
For use under harsh conditions such as mechanized washing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS125099751 L
FIS02-555-25A150 mL
FIS02-555-25B250 mL
FIS02-555-25D600 mL
FIS02-555-25F1 L
FIS02-555-25H2000 mL
FKS119115162000 mL
FIS02-555-25K4000 mL
Catalog number FKS12509975
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1 L
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes.
Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot.
Specifications
Capacity (English)33.81 oz.
Diameter (English) Outer3.96 in.
Diameter (Metric) Outer10.8 cm
Capacity (Metric)1 L
MaterialBorosilicate Glass
For Use With (Application)Provides The Best Mechanical Strength Under Harsh Conditions Such as Mechanized Washings
Graduation Range50 to 1000 mL
Height (English)6.14 in.
Height (Metric)15.6 cm
Increments50 mL
Label TypeExtra Large Marking
Quantity6 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960
ScaleDual Graduated Metric Scale
StyleGriffin
Unit SizePK