Polypropylene 'T' Shaped Untapered Connectors
Polypropylene 'T' Shaped Untapered Connectors
Actual product may vary
Fisherbrand
Polypropylene 'T' Shaped Untapered Connectors
The ideal leak-proof solution for connecting tubes. Kartell™ Polypropylene 'T' Shaped Untapered Connectors are useful for connecting tubes of various diameters.
Have Questions?
Catalog NumberDiameter (Metric) Inner
FKS125097668.2 mm Bore
Catalog number FKS12509766
Price (SGD)
36.00
PK
Add to cart
Diameter (Metric) Inner:
8.2 mm Bore
Price (SGD)
36.00
PK
Add to cart
  • Excellent chemical and temperature resistance
  • With serrations to hold hoses tightly and prevent slipping
  • Autoclavable
  • Untapered
Specifications
Certifications/ComplianceCE IVD
Diameter (Metric)12 mm
AutoclavableAutoclavable
Diameter (Metric) Inner8.2 mm Bore
MaterialPolypropylene
Packaging Quantity10 pcs.
Tubing Size12 x 8.2mm
Port Size8.2mm Bore
Sterilization MethodAutoclave
ShapeT
ColorNatural
Product TypeT Untapered Connector
Unit SizePK