1x1 image pixel for data collection
Quickfit™ Borosilicate Glass Air Condenser
Product Image
Fisherbrand
Quickfit™ Borosilicate Glass Air Condenser
Manufactured from Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Air Condenser creates condensation of materials with boiling points above 150°C.
Have Questions?
Catalog number FKS12508845
Price (SGD)
104.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Price (SGD)
104.00
EA
Add to cart
  • Manufactured from Pyrex™ Borosilicate Glass
  • Simple condensers for condensation of materials with boiling points above 150°C
Specifications
For Use With (Application)For condensation of materials with boiling points above 150°C
Length (Metric)260 mm
Quantity1 Pack
Standard Taper Joint No14/23
MaterialBorosilicate Glass
TypeAir Condenser
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link