Aluminum Labjacks
Aluminum Labjacks
Actual product may vary
Aluminum Labjacks
Aluminum Labjacks
Aluminum Labjacks
Fisherbrand
Aluminum Labjacks
Provides for precise height adjustment with the turn of a screw. Fisherbrand™ Aluminum Labjacks are manufactured to close tolerances with a stainless steel operating screw.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Length (Metric) BaseWidth (Metric) Base
FKS1248406057 to 290 mm200 mm200 mm
FKS1182509160 to 290 mm150 mm150 mm
Catalog number FKS12484060
Price (SGD)
672.00
EA
Add to cart
Height (Metric):
57 to 290 mm
Length (Metric) Base:
200 mm
Width (Metric) Base:
200 mm
Price (SGD)
672.00
EA
Add to cart
  • Blue anodized finish
  • Completely stable and free from rocking movements
  • Top and bottom plates are specially designed aluminum extrusion giving exceptional rigid and robust support
Specifications
MaterialAluminum
Height (Metric)57 to 290 mm
Length (Metric) Base200 mm
Width (Metric) Base200 mm
IncludesWith stainless steel operating screw
Product TypeLab Jack
Quantity1 Ea.
Unit SizeEA