Grade 1573 Filter Paper
Grade 1573 Filter Paper
Actual product may vary
Fisherbrand
Grade 1573 Filter Paper
High wet strength makes it easy to scrape or wash off precipitate. Whatman™ Grade 1573 Filter Paper is an extremely strong, smooth-surface filter paper with a high wet strength due to the addition of a small quantity of chemically stable resin.
Have Questions?
Catalog NumberDimensions (L x W)
FKS1247429270 mm x 80 m
Catalog number FKS12474292
Price (SGD)
243.00
EA
Add to cart
Dimensions (L x W):
70 mm x 80 m
Price (SGD)
243.00
EA
Add to cart
  • Normal qualitative applications will not introduce any significant impurities into the filtrate.
  • The resins contain nitrogen, so these grades should not be used in Kjeldahl estimations.
  • Resistant against: sulfuric and nitric acid solutions (up to 40% at 50°C); hydrochloric (up to 10% at 100°C, 20% at 60°C, 25% at 20°C); and alkalis (up to 10% at 20°C).
Specifications
Basis Weight88 g/m2
DescriptionGrade 1573 Qualitative Filter Paper Wet-Strengthened, roll, 70 mm x 80 m
FormatRoll
MaterialResin-infused Cellulose
Particle Retention25 μm
Filtration SpeedApprox. 25 s
For Use With (Application)A fast filter paper with high wet strength. It has a very smooth surface making it easy to scrape or wash off precipitate.
Grade1573
Quantity1 Ea.
Thickness (Metric)170 μm
TypeQualitative Filter Paper
Dimensions (L x W)70 mm x 80 m
Length (Metric)80m
Width (Metric)70 mm
Unit SizeEA