Stainless Steel Surgical Scalpel Handle 3G
Stainless Steel Surgical Scalpel Handle 3G
Actual product may vary
Fisherbrand
Stainless Steel Surgical Scalpel Handle 3G
Individually wrapped. Swann Morton Stainless Steel Surgical Scapel Handles contain graduations on one side to assist with training, marking of surgical site or trauma injury investigation.
Have Questions?
Catalog NumberSize
FKS12464070# 3
Catalog number FKS12464070
Price (SGD)
-
Size:
# 3
Individually wrapped. Swann Morton Stainless Steel Surgical Scapel Handles contain graduations on one side to assist with training, marking of surgical site or trauma injury investigation.
  • Stainless Steel
  • Individually wrapped
Specifications
Blade MaterialStainless Steel
Product TypeScalpel Handle
Size# 3