MBL™ Class B Autozero Pinch Clip Burette
MBL™ Class B Autozero Pinch Clip Burette
Actual product may vary
Fisherbrand
MBL™ Class B Autozero Pinch Clip Burette
Manufactured from borosilicate glass and complies with ISO 385 standards. MBL™ Class B Autozero Pinch Clip Buret provides easy filling and automatic zero. Squeeze the polyethylene bottle to fill buret, release pressure to allow excess liquid to
Have Questions?
Catalog NumberCapacity (Metric)
FKS1246155720 mL
Catalog number FKS12461557
Price (SGD)
432.00
EA
Add to cart
Capacity (Metric):
20 mL
Price (SGD)
432.00
EA
Add to cart
Manufactured from borosilicate glass and complies with ISO 385 standards. MBL™ Class B Autozero Pinch Clip Buret provides easy filling and automatic zero. Squeeze the polyethylene bottle to fill buret, release pressure to allow excess liquid to
  • Complies with ISO 385
  • Easy filling and automatic zero - Squeeze the polyethylene bottle to fill burette - Release pressure to allow excess liquid to drain back into bottle and set the burette to zero
  • Easy to read blue enamel graduations
  • Manufactured from borosilicate glass
  • With simple rubber tube and pinch valve for low cost replacement
Specifications
Capacity (Metric)20 mL
MaterialBorosilicate Glass
Graduations0.1 mL
For Use With (Application)Water Testing
IncludesJet and Polyethylene Reservoir Bottle, Pinch Clip, Rubber Tube
ScaleBlue
Tolerance±0.1 mL
TypeAutomatic Buret
ClassClass B
Certifications/ComplianceISO 385
Unit SizeEA