Initial™ Green Spirit-filled Partial Immersion Thermometer
Initial™ Green Spirit-filled Partial Immersion Thermometer
Actual product may vary
Fisher Catalogue
Initial™ Green Spirit-filled Partial Immersion Thermometer
Provides vibrant, easy to read measurements. Brannan™ Initial™ Green Spirit-filled Partial Immersion Thermometer is designed with moulded non-roll glass device to helps prevent breakage. Includes integral bulb with reinforced stirring tip.
Have Questions?
Catalog NumberLength (Metric)Weight (Metric)
FKS12444080200 mm15.8 g
Catalog number FKS12444080
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
200 mm
Weight (Metric):
15.8 g
Provides vibrant, easy to read measurements. Brannan™ Initial™ Green Spirit-filled Partial Immersion Thermometer is designed with moulded non-roll glass device to helps prevent breakage. Includes integral bulb with reinforced stirring tip.
  • Green Kerosene spirit filling is miscible in water
  • Green, miscible fluid provides vibrant, easy to read measurements
  • Moulded non-roll glass device helps to prevent breakage
  • Integral bulb with reinforced stirring tip
  • Recyclable
  • Ideal for educational use
Specifications
For Use With (Application)Ideal for educational use
Immersion TypePartial Immersion
Length (Metric)200 mm
MaterialGlass
Thermometer FillGreen Spirit
Weight (Metric)15.8 g
Product TypeLaboratory Thermometer
Max. Temperature (Metric)110°C
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +110°C
Accuracy±1.5 at 35°C
Subdivision1°C
Immersion Length76 mm
Unit SizeEA