Check Order Status
Quickfit™ Borosilicate Glass Distillation Apparatus
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Distillation Apparatus
For the purpose of separating and/or purifying components of a liquid mixture.
Have Questions?
Catalog Number
Type
FKS12443601
Distillation apparatus
Catalog number
FKS12443601
Price (SGD)
670.00
EA
Add to cart
Type:
Distillation apparatus
Price (SGD)
670.00
EA
Add to cart
Comprises Liebig condenser, stillhead and receiver adapter in a single piece
Angle and position pre-defined
Simple to set up with fewer clamps required
Less grease contamination
No air leakage under vacuum
No liquid hold-up at the joints
Specifications
Length (Metric)
Approx. effective condenser length: 155mm
Type
Distillation apparatus
Material
Borosilicate Glass
Standard Taper Joint
Cone size: 19/26; Socket size: 14/23