27BU Organic Chemistry Kit
27BU Organic Chemistry Kit
Actual product may vary
Fisherbrand
27BU Organic Chemistry Kit
Ideal organic chemistry kit for use in schools, colleges, and universities. Quickfit™ 27BU Organic Chemistry Kit is perfect for introducing students to the principal techniques of organic chemistry.
Have Questions?
Catalog number FKS12433461
Price (SGD)
1,547.00
EA
Add to cart
Price (SGD)
1,547.00
EA
Add to cart
Ideal organic chemistry kit for use in schools, colleges, and universities. Quickfit™ 27BU Organic Chemistry Kit is perfect for introducing students to the principal techniques of organic chemistry.
  • The most popular Quickfit™ set for schools, colleges and universities
  • Comprises nine basic items of Quickfit™ glassware all with size 14/23 ground glass joints for compatibility
  • Allows the construction of at least 8 assemblies including steam distillation, reflux with addition and preparation
  • Scale based on starting material quantities up to 30g
  • Housed in a robust cardboard box with foam rubber liner to provide safe storage and transit
  • Supplied with a CD-ROM which includes practical details of some typical assemblies
Specifications
IncludesPear shaped flask, Stillhead, Liebig condenser, Screwcap adapter, Receiver adapter, Air leak/steam inlet tube, Dropping funnel with GP Rotoflo tap, Stopper, Thermometer
Product TypeOrganic Chemistry Kit
Unit SizeEA