Use these wash bottles for a variety of laboratory applications. S Murray™ Wash Bottles with Hand Triggers feature an adjustable nozzle which produces a powerful intermittent or continuous jet or fine spray.
Have Questions?
Catalog Number
Capacity (Metric)
FKS12413070
500 mL
Catalog number FKS12413070
Price (SGD)
10.00
EA
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
10.00
EA
Add to cart
Head is mounted on a polyethylene bottle shaped to fit the hand